Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmem243
Entrez Gene ID
780238
Gene Name
transmembrane protein 243
Gene Aliases
c7orf23
Chromosome Location
Chr.3: 71607481 - 71618128 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079310.1 NP_001072778.1 2 - 3 684 92
XM_012959285.2 XP_012814739.1 3 - 4 480 92
GenBank mRNA BC118807.1 - 2 - 3 684 92
CT025493.2 - 2 - 3 145 92
CU075814.1 - 3 - 4 393 92

View Full Product Details