Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vstm2b
Entrez Gene ID
780250
Gene Name
V-set and transmembrane domain containing 2B
Gene Aliases
-
Chromosome Location
Chr.4: 23426852 - 23485916 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079321.1 NP_001072789.1 1 - 2 664 91
XM_012961411.2 XP_012816865.1 2 - 3 872 91
GenBank mRNA BC121271.1 - 1 - 2 664 91
CU025205.1 - 1 - 2 469 91

View Full Product Details