Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ogfrl1
Entrez Gene ID
100037841
Gene Name
opioid growth factor receptor-like 1
Gene Aliases
-
Chromosome Location
Chr.5: 91584603 - 91616693 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097287.1 NP_001090756.1 2 - 3 375 87
XM_018093724.1 XP_017949213.1 2 - 3 176 87
GenBank mRNA BC128651.1 - 2 - 3 375 87

View Full Product Details