Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sst
Entrez Gene ID
100037904
Gene Name
somatostatin
Gene Aliases
smst, som
Chromosome Location
Chr.5: 100658319 - 100659858 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097338.1 NP_001090807.1 1 - 2 201 72
GenBank mRNA BC135242.1 - 1 - 2 201 72

View Full Product Details