Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
igfbp2
Entrez Gene ID
100101720
Gene Name
insulin like growth factor binding protein 2
Gene Aliases
IGFBP-2, ibp2, igf-bp53
Chromosome Location
Chr.9: 80084239 - 80116588 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100237.1 NP_001093707.1 3 - 4 1021 80
GenBank mRNA BC135928.1 - 3 - 4 1021 80

View Full Product Details