Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
dlx1
Entrez Gene ID
100101750
Gene Name
distal-less homeobox 1
Gene Aliases
-
Chromosome Location
Chr.9: 62320946 - 62327265 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100257.1 NP_001093727.1 - 510 86
GenBank mRNA BC127554.1 - - 510 86

View Full Product Details