Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
hdhd5
Entrez Gene ID
100101802
Gene Name
haloacid dehalogenase like hydrolase domain containing 5
Gene Aliases
cecr5
Chromosome Location
Chr.Un|NW_016683387: 521398 - 528491 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100283.1 NP_001093753.1 - 705 62
GenBank mRNA BC136143.1 - - 705 62

View Full Product Details