Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aip
Entrez Gene ID
100124770
Gene Name
aryl hydrocarbon receptor interacting protein
Gene Aliases
xaip
Chromosome Location
Chr.Un|NW_016683379: 66959 - 140839 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102749.1 NP_001096219.1 1 - 2 110 90
XM_012965878.2 XP_012821332.1 1 - 2 215 90
GenBank mRNA BC158230.1 - 1 - 2 108 90
CR762165.2 - 1 - 2 110 90

View Full Product Details