Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nptx2
Entrez Gene ID
100124882
Gene Name
neuronal pentraxin 2
Gene Aliases
narp, np-ii, np2
Chromosome Location
Chr.9: 20329064 - 20344099 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102834.1 NP_001096304.1 4 - 5 1350 83
GenBank mRNA BC135649.1 - 4 - 5 1350 83

View Full Product Details