Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lmcd1
Entrez Gene ID
100124907
Gene Name
LIM and cysteine-rich domains 1
Gene Aliases
-
Chromosome Location
Chr.4: 120664986 - 120688183 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102853.1 NP_001096323.1 3 - 4 428 99
XM_012960465.2 XP_012815919.1 3 - 4 454 99
GenBank mRNA BC135721.1 - 3 - 4 428 99

View Full Product Details