Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
med28
Entrez Gene ID
100125095
Gene Name
mediator complex subunit 28
Gene Aliases
-
Chromosome Location
Chr.1: 16439238 - 16442155 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103006.1 NP_001096476.1 1 - 2 179 101
GenBank mRNA BC136218.1 - 1 - 2 179 101
BC155038.1 - 1 - 2 159 101
CT010556.2 - 1 - 2 171 101

View Full Product Details