Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gipc3
Entrez Gene ID
100125129
Gene Name
GIPC PDZ domain containing family member 3
Gene Aliases
-
Chromosome Location
Chr.1: 74638455 - 74660952 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103032.1 NP_001096502.1 4 - 5 738 71
GenBank mRNA BC135287.1 - 4 - 5 738 71

View Full Product Details