Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
wdr49
Entrez Gene ID
549615
Gene Name
WD repeat domain 49
Gene Aliases
-
Chromosome Location
Chr.5: 101534861 - 101577580 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272027.1 NP_001258956.1 17 - 18 3189 96
XM_012963038.2 XP_012818492.1 18 - 19 3405 96
XM_012963039.2 XP_012818493.1 18 - 19 3462 96
GenBank mRNA BC121341.1 - 13 - 14 1980 96
BC161191.1 - 17 - 18 3189 96

View Full Product Details