Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rsbn1
Entrez Gene ID
100127593
Gene Name
round spermatid basic protein 1
Gene Aliases
-
Chromosome Location
Chr.2: 75797398 - 75822557 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112954.2 NP_001106425.2 2 - 3 1146 90
GenBank mRNA BC153367.1 - 2 - 3 1113 90
BX752862.1 - 5 - 6 776 90

View Full Product Details