Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp2c18
Entrez Gene ID
100127628
Gene Name
cytochrome P450 family 2 subfamily C member 18
Gene Aliases
cpci, cyp2c, cyp2c17, p450-6b/29c, p450iic17
Chromosome Location
Chr.8: 57738816 - 57756570 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112980.1 NP_001106451.1 7 - 8 1163 98
GenBank mRNA BC153744.1 - 7 - 8 1163 98

View Full Product Details