Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rell1
Entrez Gene ID
100127768
Gene Name
RELT-like 1
Gene Aliases
-
Chromosome Location
Chr.1: 25920301 - 25940305 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113093.2 NP_001106564.1 - 389 74
GenBank mRNA BC155011.1 - - 339 74
EL821376.1 - 2 - 3 389 74
EL836327.1 - 2 - 3 389 74

View Full Product Details