Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cep19
Entrez Gene ID
100135164
Gene Name
centrosomal protein 19kDa
Gene Aliases
c3orf34
Chromosome Location
Chr.5: 129897151 - 129903645 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113870.1 NP_001107342.1 2 - 3 190 106
GenBank mRNA BC157153.1 - 2 - 3 190 106
BC171127.1 - 2 - 3 171 106
BC171129.1 - 2 - 3 171 106

View Full Product Details