Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rras
Entrez Gene ID
100135709
Gene Name
RAS related
Gene Aliases
r-ras
Chromosome Location
Chr.7: 102860140 - 102886506 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114248.1 NP_001107720.1 5 - 6 561 90
GenBank mRNA BC158407.1 - 5 - 6 561 90

View Full Product Details