Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dimt1
Entrez Gene ID
779588
Gene Name
DIM1 dimethyladenosine transferase 1 homolog
Gene Aliases
dim1, dimt1l, hussy5
Chromosome Location
Chr.1: 176047272 - 176055592 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122722.1 NP_001116194.1 11 - 12 913 87
GenBank mRNA BC123077.1 - 11 - 12 869 87
BC160444.1 - 11 - 12 913 87
BC169134.1 - 11 - 12 916 87
CU075813.1 - 11 - 12 874 87

View Full Product Details