Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dimt1
Entrez Gene ID
779588
Gene Name
DIM1 dimethyladenosine transferase 1 homolog
Gene Aliases
dim1, dimt1l, hussy5
Chromosome Location
Chr.1: 176047272 - 176055592 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122722.1 NP_001116194.1 3 - 4 261 83
GenBank mRNA BC123077.1 - 3 - 4 217 83
BC160444.1 - 3 - 4 261 83
BC169134.1 - 3 - 4 264 83
CU075813.1 - 3 - 4 222 83

View Full Product Details