Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
timm50
Entrez Gene ID
100145004
Gene Name
translocase of inner mitochondrial membrane 50 homolog
Gene Aliases
-
Chromosome Location
Chr.8: 60595891 - 60614458 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126566.1 NP_001120038.1 4 - 5 357 92
GenBank mRNA BC158346.1 - 4 - 5 357 92
CT030353.1 - 4 - 5 398 92

View Full Product Details