Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
timm50
Entrez Gene ID
100145004
Gene Name
translocase of inner mitochondrial membrane 50 homolog
Gene Aliases
-
Chromosome Location
Chr.8: 60595891 - 60614458 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126566.1 NP_001120038.1 8 - 9 738 99
GenBank mRNA BC158346.1 - 8 - 9 738 99
CT030353.1 - 8 - 9 779 99

View Full Product Details