Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
utp3
Entrez Gene ID
100145025
Gene Name
UTP3, small subunit processome component
Gene Aliases
CRL1, CRLZ1, SAS10
Chromosome Location
Chr.7: 77972498 - 77990897 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126577.1 NP_001120049.1 5 - 6 335 73
XM_012971526.1 XP_012826980.1 5 - 6 436 73
GenBank mRNA BC158387.1 - 5 - 6 335 73

View Full Product Details