Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tekt4
Entrez Gene ID
100145069
Gene Name
tektin 4
Gene Aliases
-
Chromosome Location
Chr.9: 13406637 - 13414469 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126596.1 NP_001120068.1 1 - 2 544 88
GenBank mRNA BC158484.1 - 1 - 2 544 88

View Full Product Details