Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
jph1
Entrez Gene ID
100145273
Gene Name
junctophilin 1
Gene Aliases
-
Chromosome Location
Chr.6: 106597943 - 106651104 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126752.1 NP_001120224.1 2 - 3 1390 77
GenBank mRNA BC160387.1 - 2 - 3 1390 77

View Full Product Details