Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clhc1
Entrez Gene ID
100145484
Gene Name
clathrin heavy chain linker domain containing 1
Gene Aliases
c2orf63
Chromosome Location
Chr.5: 20222297 - 20236576 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126935.1 NP_001120407.1 11 - 12 1662 90
XM_012962732.2 XP_012818186.1 11 - 12 1742 90
XM_012962733.2 XP_012818187.1 11 - 12 1808 90
XM_012962734.2 XP_012818188.1 11 - 12 1706 90
XM_012962735.2 XP_012818189.1 12 - 13 2213 90
XM_012962736.2 XP_012818190.1 12 - 13 2210 90
XM_012962737.1 XP_012818191.1 11 - 12 1699 90
XM_012962738.2 XP_012818192.2 9 - 10 1275 90
GenBank mRNA BC161123.1 - 11 - 12 1662 90

View Full Product Details