Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gbp6
Entrez Gene ID
100145795
Gene Name
guanylate binding protein family member 6
Gene Aliases
-
Chromosome Location
Chr.7: 108988381 - 109005231 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127156.1 NP_001120628.1 10 - 11 1812 80
XM_018095277.1 XP_017950766.1 10 - 11 1855 80
GenBank mRNA BC161780.1 - 10 - 11 1812 80

View Full Product Details