Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vps18
Entrez Gene ID
100158548
Gene Name
VPS18, CORVET/HOPS core subunit
Gene Aliases
-
Chromosome Location
Chr.8: 95019104 - 95030294 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127982.1 NP_001121454.1 2 - 3 240 78
XM_018096166.1 XP_017951655.1 1 - 2 96 78
GenBank mRNA BC166213.1 - 2 - 3 240 78

View Full Product Details