Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ctc1
Entrez Gene ID
100158579
Gene Name
CTS telomere maintenance complex component 1
Gene Aliases
aaf-132, aaf132, c17orf68
Chromosome Location
Chr.3: 131669488 - 131680412 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128009.1 NP_001121481.1 - 1725 95
GenBank mRNA BC166255.1 - - 1725 95

View Full Product Details