Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
phf5a
Entrez Gene ID
100170460
Gene Name
PHD finger protein 5A
Gene Aliases
-
Chromosome Location
Chr.4: 109952721 - 109958229 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130240.1 NP_001123712.1 1 - 2 182 96
GenBank mRNA BC167322.1 - 1 - 2 182 96
CR942721.2 - 1 - 2 61 96

View Full Product Details