Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
emx2
Entrez Gene ID
100038069
Gene Name
empty spiracles homeobox 2
Gene Aliases
emx-2, xemx2
Chromosome Location
Chr.Un|NW_016683776: 56781 - 64900 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134812.1 NP_001128284.1 1 - 2 700 67
GenBank mRNA BC169182.1 - 1 - 2 700 67
CT030607.1 - 1 - 2 680 67

View Full Product Details