Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
c16orf45
Entrez Gene ID
100216095
Gene Name
chromosome 16 open reading frame 45
Gene Aliases
-
Chromosome Location
Chr.9: 28218153 - 28338965 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142078.1 NP_001135550.1 1 - 2 132 61
GenBank mRNA BC168030.1 - 1 - 2 132 61

View Full Product Details