Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fdft1
Entrez Gene ID
100216201
Gene Name
farnesyl-diphosphate farnesyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683369: 821041 - 840646 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142170.3 NP_001135642.3 2 - 3 210 63
GenBank mRNA BC168496.1 - 2 - 3 210 63

View Full Product Details