Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dlx6
Entrez Gene ID
100216239
Gene Name
distal-less homeobox 6
Gene Aliases
dlx-6, dlx6-a, dlx6-b
Chromosome Location
Chr.Un|NW_016683410: 1007769 - 1012729 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142206.1 NP_001135678.1 1 - 2 2641 67
GenBank mRNA BC168605.1 - 1 - 2 2641 67

View Full Product Details