Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
asn2
Entrez Gene ID
100192349
Gene Name
asparagine synthetase2
Gene Aliases
AsnS2, ZEAMMB73_149260
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001137541.1 NP_001131013.1 - 1469 74
GenBank mRNA BT063342.1 - - 1490 74
BT064177.1 - - 1567 74
BT083882.1 - - 1490 74
FM211452.1 - - 1465 74
FM212634.1 - - 1469 74

View Full Product Details