Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LOC100285243
Entrez Gene ID
100285243
Gene Name
ATMAP70-2
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001158137.1 NP_001151609.1 - 280 75
GenBank mRNA EU971391.1 - - 280 75

View Full Product Details