Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
AGP2
Entrez Gene ID
542737
Gene Name
ADP-glucose pyrophosphorylase
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112247.1 NP_001105717.1 - 1017 85
GenBank mRNA Z38111.1 - - 1017 85

View Full Product Details