Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
p5cr
Entrez Gene ID
606449
Gene Name
pyrroline 5-carboxylate reductase
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112317.1 NP_001105787.1 - 527 61
GenBank mRNA BT037020.1 - - 527 61
DQ026301.1 - - 527 61
EU968742.1 - - 539 61

View Full Product Details