Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LOX2
Entrez Gene ID
100037802
Gene Name
lipoxygenase
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112503.1 NP_001105973.1 - 1890 80
GenBank mRNA DQ335761.1 - - 1890 80

View Full Product Details