Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LOC100193210
Entrez Gene ID
100193210
Gene Name
ATARP7
Gene Aliases
ZEAMMB73_095268
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001138363.1 NP_001131835.1 - 297 89
GenBank mRNA BT035416.1 - - 297 89

View Full Product Details