Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
pcna1
Entrez Gene ID
542425
Gene Name
proliferating cell nuclear antigen1
Gene Aliases
PCNA
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111991.1 NP_001105461.1 - 1051 90
GenBank mRNA AY110234.1 - - 1051 90
BT055362.1 - - 1045 90
X79065.1 - - 1051 90

View Full Product Details