Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
lac2
Entrez Gene ID
732786
Gene Name
laccase-like
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112404.1 NP_001105874.1 - 1250 96
GenBank mRNA AM086214.1 - - 1250 96

View Full Product Details