Gene Transcripts
- Gene Symbol
- Gapdh-ps1
- Entrez Gene ID
- 366057
- Gene Name
- glyceraldehyde-3-phosphate dehydrogenase, pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 44736647 - 44738545 on Build Rnor_6.0
- UniGene ID
- Rn.160981
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003722.1 |
|
1 |
35 |
|
GenBank mRNA |
BC087069.1 |
- |
1 |
35 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CK598450.1 |
- |
|
34 |
|
Chr. - - [-] |
View Full Product Details