Gene Transcripts
- Gene Symbol
- Obox2
- Entrez Gene ID
- 246792
- Gene Name
- oocyte specific homeobox 2
- Gene Aliases
- Chromosome Location
- Chr. 7: 15388851 - 15398545 on Build GRCm38
- UniGene ID
- Mm.26391
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC094912.1 |
- |
4 |
1195 |
|
DV647958.1 |
- |
1 |
703 |
|
- Gene Symbol
- Obox1
- Entrez Gene ID
- 71468
- Gene Name
- oocyte specific homeobox 1
- Gene Aliases
- 7420700M11Rik, AU015708, AU024152
- Chromosome Location
- Chr. 7: 15547257 - 15556846 on Build GRCm38
- UniGene ID
- Mm.415195
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK136061.1 |
- |
4 |
1213 |
|
View Full Product Details