Gene Transcripts
- Gene Symbol
- Snora7a
- Entrez Gene ID
- 100217451
- Gene Name
- small nucleolar RNA, H/ACA box 7A
- Gene Aliases
- MBI-141
- Chromosome Location
- Chr. 6: 115807975 - 115808103 on Build GRCm38
- UniGene ID
- Mm.484708
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028546.1 |
|
1 |
18 |
|
GenBank mRNA |
AF357398.1 |
- |
1 |
18 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DT918153.1 |
- |
1 |
263 |
|
Chr. 6 - 115807111 - 115808348 [-] |
View Full Product Details