Gene Transcripts

Gene Symbol
Vmn2r-ps11
Entrez Gene ID
319210
Gene Name
vomeronasal 2, receptor, pseudogene 11
Gene Aliases
4930518C23Rik
Chromosome Location
Chr. 3: 64632863 - 64844743 on Build GRCm38
UniGene ID
Mm.246646
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003962.1 3 322
GenBank mRNA AK029734.1 - 3 323
BB615497.1 - 3 323

Gene Symbol
Vmn2r7
Entrez Gene ID
319217
Gene Name
vomeronasal 2, receptor 7
Gene Aliases
4933425M15Rik
Chromosome Location
Chr. 3: 64690660 - 64719602 on Build GRCm38
UniGene ID
Mm.246646
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK030224.1 - 1055

View Full Product Details