Gene Transcripts
- Gene Symbol
- Snora15
- Entrez Gene ID
- 100113364
- Gene Name
- small nucleolar RNA, H/ACA box 15
- Gene Aliases
- MBI-79
- Chromosome Location
- Chr. 5: 129794561 - 129794676 on Build GRCm38
- UniGene ID
- Mm.449738
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003681.1 |
|
|
11 |
|
GenBank mRNA |
AF357410.1 |
- |
|
11 |
|
View Full Product Details