Gene Transcripts

Gene Symbol
AKAP17A
Entrez Gene ID
8227
Gene Name
A-kinase anchoring protein 17A
Gene Aliases
721P, AKAP-17A, CCDC133, CXYorf3, DXYS155E, PRKA17A, SFRS17A, XE7, XE7Y
Chromosome Location
Chr. Y: 1591593 - 1602520 on Build GRCh38
UniGene ID
Hs.522572
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027383.1 5 1380
GenBank mRNA BC028151.1 - 4 2053
L03426.1 - 5 1350


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AL045946.1 - 2 62 Chr. X - 1599403 - 1601063 [+]

View Full Product Details