Gene Transcripts
- Gene Symbol
- CETN4P
- Entrez Gene ID
- 729338
- Gene Name
- centrin 4, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 4: 122730189 - 122732458 on Build GRCh38
- UniGene ID
- Hs.647968
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024041.1 |
|
3 |
470 |
|
GenBank mRNA |
AV733354.1 |
- |
3 |
470 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ429158.1 |
- |
3 |
563 |
|
Chr. 4 - 122730161 - 122732419 [-] |
GenBankmRNA |
HY015149.1 |
- |
3 |
492 |
|
Chr. 4 - 122730300 - 122732697 [-] |
View Full Product Details