Gene Transcripts
- Gene Symbol
- HAUS1
- Entrez Gene ID
- 115106
- Gene Name
- HAUS augmin like complex subunit 1
- Gene Aliases
- CCDC5, HEI-C, HEIC, HsT1461
- Chromosome Location
- Chr. 18: 46104332 - 46128333 on Build GRCh38
- UniGene ID
- Hs.436617
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026978.1 |
|
2, 3 |
327 |
|
GenBank mRNA |
BC005958.1 |
- |
2, 3 |
263 |
|
View Full Product Details